Assay Details
| Gene Symbol: Arhgef38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK052594.1 | - | 380 | 70 | ||
| AK052650.1 | - | 138 | 70 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgef38| Gene Symbol: Arhgef38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK052594.1 | - | 380 | 70 | ||
| AK052650.1 | - | 138 | 70 | |||
Set Membership: |
|