Assay Details
| Gene Symbol: Arhgef38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK052594.1 | - | 619 | 87 | ||
| AK052650.1 | - | 377 | 87 | |||
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2327577 | AK052650.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P108733 | AK052594.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596781 | AK021399.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 182689 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 316570 | AK015558.1 | genbank_mrna_mouse |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171584 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2327577 | AK052650.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05079028 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171584 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00760708 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05079012 | - | - |
| Whole Mouse Genome G4122A | A_52_P285207 | AK052594.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P318710 | AK052650.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171584 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02608294 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541309 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541274 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541318 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03094406 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02375328 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541267 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541265 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541307 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541321 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02608289 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541310 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541252 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541312 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02608286 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541237 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541338 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541224 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541276 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02371001 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541319 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541258 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541285 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541257 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541281 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541316 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03094398 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541236 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02510391 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541328 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541320 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541303 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541333 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541268 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541317 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02287855 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541306 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541238 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02823073 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541330 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541325 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541326 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02608291 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541270 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541336 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02608288 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541340 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02370987 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541324 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541256 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541249 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02510392 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03187152 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541260 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541284 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02375326 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541301 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541343 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541291 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541331 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02630699 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03057515 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02864237 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02697595 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541298 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02371006 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22541314 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 101450546 | - | - |

Arhgef38
Genomic Map