Assay Details
| Gene Symbol: Slc22a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011395.2 | NP_035525.1 | 3 - 4 | 1055 | 83 | |
| GenBank mRNA | AF078750.1 | - | 3 - 4 | 1055 | 83 | |
| AF082566.1 | - | 3 - 4 | 938 | 83 | ||
| AK143988.1 | - | 3 - 4 | 725 | 83 | ||
| BC137661.1 | - | 3 - 4 | 930 | 8860680 | 83 | |
| BC137662.1 | - | 3 - 4 | 930 | 8860681 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc22a3
Genomic Map