Assay Details
| Gene Symbol: Plg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008877.3 | NP_032903.3 | 10 - 11 | 1314 | 72 | |
| GenBank mRNA | AK132254.1 | - | 10 - 11 | 1283 | 72 | |
| AY424276.1 | - | 10 - 11 | 1282 | 72 | ||
| BC014773.1 | - | 10 - 11 | 1315 | 4193957 | 72 | |
| BC057186.1 | - | 10 - 11 | 1289 | 5097123 | 72 | |
| J04766.1 | - | 10 - 11 | 1279 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plg
Genomic Map