Assay Details
| Gene Symbol: Plg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008877.3 | NP_032903.3 | 16 - 17 | 2086 | 61 | |
| GenBank mRNA | AK132254.1 | - | 16 - 17 | 2055 | 61 | |
| BC014773.1 | - | 16 - 17 | 2087 | 4193957 | 61 | |
| BC057186.1 | - | 16 - 17 | 2061 | 5097123 | 61 | |
| J04766.1 | - | 16 - 17 | 2051 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plg
Genomic Map