Assay Details
| Gene Symbol: Plg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008877.3 | NP_032903.3 | 2 - 3 | 246 | 75 | |
| GenBank mRNA | AK040840.1 | - | 2 - 3 | 197 | 75 | |
| AK132254.1 | - | 2 - 3 | 215 | 75 | ||
| AY424276.1 | - | 2 - 3 | 214 | 75 | ||
| BC014773.1 | - | 2 - 3 | 247 | 4193957 | 75 | |
| BC057186.1 | - | 2 - 3 | 221 | 5097123 | 75 | |
| BY131661.1 | - | 2 - 3 | 246 | 75 | ||
| J04766.1 | - | 2 - 3 | 211 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plg
Genomic Map