Assay Details
| Gene Symbol: Pnldc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034866.1 | NP_001030038.1 | 12 - 13 | 1077 | 77 | |
| GenBank mRNA | BC158033.1 | - | 12 - 13 | 1077 | 9007336 | 77 |
| BC172002.1 | - | 12 - 13 | 1080 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pnldc1
Genomic Map