Assay Details
| Gene Symbol: Mas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008552.4 | NP_032578.2 | 2 - 3 | 159 | 151 | |
| GenBank mRNA | AK015036.1 | - | 2 - 3 | 150 | 151 | |
| AK030261.1 | - | 2 - 3 | 149 | 151 | ||
| AK030265.1 | - | 2 - 3 | 160 | 151 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK015036.1, AK030261.1, AK030265.1, NM_008552.4 transcripts.
Set Membership: |
|

Mas1
Genomic Map