Assay Details
| Gene Symbol: Mas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008552.4 | NP_032578.2 | 4 - 5 | 454 | 86 | |
| GenBank mRNA | AK015036.1 | - | 4 - 5 | 445 | 86 | |
| AK030261.1 | - | 4 - 5 | 444 | 86 | ||
| AK030265.1 | - | 4 - 5 | 455 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK015036.1,AK030261.1,AK030265.1,NM_008552.4 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mas1
Genomic Map