Assay Details
| Gene Symbol: Mtmr9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177594.1 | NP_808262.1 | 3 - 4 | 729 | 109 | |
| GenBank mRNA | AK035026.1 | - | 3 - 4 | 744 | 109 | |
| AK044957.1 | - | 3 - 4 | 759 | 109 | ||
| AK152320.1 | - | 3 - 4 | 515 | 109 | ||
| BC046275.1 | - | 3 - 4 | 729 | 6307469 | 109 | |
| BC145946.1 | - | 3 - 4 | 484 | 40131249 | 109 | |
| BC145948.1 | - | 3 - 4 | 484 | 40131251 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mtmr9
Genomic Map