Assay Details
| Gene Symbol: Tdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021480.5 | NP_067455.5 | 1 - 2 | 384 | 67 | |
| GenBank mRNA | AF134346.2 | - | 1 - 2 | 48 | 67 | |
| AK077571.1 | - | 1 - 2 | 59 | 67 | ||
| BC058860.1 | - | 1 - 2 | 399 | 30014660 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF134346.2, AK077571.1, BC058860.1, NM_021480.5 transcripts.
Set Membership: |
|

Tdh
Genomic Map