Assay Details
| Gene Symbol: Tdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021480.5 | NP_067455.5 | 8 - 9 | 1386 | 80 | |
| GenBank mRNA | AF134346.2 | - | 8 - 9 | 1050 | 80 | |
| AK077571.1 | - | 8 - 9 | 1061 | 80 | ||
| AY116662.1 | - | 7 - 8 | 987 | 80 | ||
More Information
Set Membership: |
|

Tdh
Genomic Map