Assay Details
| Gene Symbol: Prss51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK005740.1 | - | 2 - 3 | 247 | 95 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK005740.1 transcripts.
Set Membership: |
|

Prss51
Genomic Map