Assay Details
| Gene Symbol: Rcbtb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027764.2 | NP_082040.2 | 4 - 5 | 850 | 63 | |
| GenBank mRNA | AK033501.1 | - | 4 - 5 | 784 | 63 | |
| AK088299.1 | - | 4 - 5 | 839 | 63 | ||
| AK089923.1 | - | 5 - 6 | 851 | 63 | ||
| BC067005.1 | - | 5 - 6 | 904 | 5721370 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK089923.1 transcripts.
Set Membership: |
|

Rcbtb1
Genomic Map