Assay Details
| Gene Symbol: Rcbtb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027764.2 | NP_082040.2 | 6 - 7 | 1106 | 86 | |
| GenBank mRNA | AK033501.1 | - | 6 - 7 | 1040 | 86 | |
| AK088299.1 | - | 6 - 7 | 1095 | 86 | ||
| AK089923.1 | - | 7 - 8 | 1107 | 86 | ||
| BC067005.1 | - | 7 - 8 | 1160 | 5721370 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rcbtb1
Genomic Map