Assay Details
| Gene Symbol: Rcbtb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027764.2 | NP_082040.2 | 7 - 8 | 1254 | 63 | |
| GenBank mRNA | AK033501.1 | - | 7 - 8 | 1188 | 63 | |
| AK088299.1 | - | 7 - 8 | 1243 | 63 | ||
| AK089923.1 | - | 8 - 9 | 1255 | 63 | ||
| BC067005.1 | - | 8 - 9 | 1308 | 5721370 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rcbtb1
Genomic Map