Assay Details
| Gene Symbol: Rcbtb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027764.2 | NP_082040.2 | 9 - 10 | 1576 | 139 | |
| GenBank mRNA | AK033501.1 | - | 9 - 10 | 1510 | 139 | |
| AK088299.1 | - | 9 - 10 | 1565 | 139 | ||
| AK089923.1 | - | 10 - 11 | 1577 | 139 | ||
| BC040764.1 | - | 2 - 3 | 271 | 139 | ||
| BC067005.1 | - | 10 - 11 | 1630 | 5721370 | 139 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rcbtb1
Genomic Map