Assay Details
| Gene Symbol: Kpna3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008466.5 | NP_032492.1 | 5 - 6 | 439 | 86 | |
| GenBank mRNA | AF020772.1 | - | 5 - 6 | 283 | 86 | |
| AK011559.1 | - | 5 - 6 | 310 | 86 | ||
| AK049373.1 | - | 5 - 6 | 310 | 86 | ||
| AK161664.1 | - | 5 - 6 | 367 | 86 | ||
| BC026885.1 | - | 5 - 6 | 333 | 3979788 | 86 | |
| CJ063714.1 | - | 5 - 6 | 524 | 86 | ||
More Information
Set Membership: |
|

Kpna3
Genomic Map