Assay Details
| Gene Symbol: Slc9b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178877.6 | NP_849208.4 | 3 - 4 | 403 | 60 | |
| GenBank mRNA | AK076505.1 | - | 4 - 5 | 444 | 60 | |
| BC090977.1 | - | 3 - 4 | 356 | 30503721 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc9b2
Genomic Map