Assay Details
| Gene Symbol: Slc9b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178877.6 | NP_849208.4 | 6 - 7 | 843 | 78 | |
| GenBank mRNA | BC090977.1 | - | 6 - 7 | 796 | 30503721 | 78 |
More Information
Set Membership: |
|

Slc9b2
Genomic Map