Assay Details
| Gene Symbol: Abcd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008991.2 | NP_033017.2 | 13 - 14 | 1251 | 114 | |
| GenBank mRNA | AK004660.1 | - | 13 - 14 | 1232 | 114 | |
| AK144060.1 | - | 13 - 14 | 1233 | 114 | ||
| AK153298.1 | - | 13 - 14 | 1209 | 114 | ||
| BC009119.1 | - | 13 - 14 | 1243 | 3154529 | 114 | |
| BC054446.1 | - | 13 - 14 | 1199 | 6477711 | 114 | |
| CB245413.1 | - | 8 - 9 | 699 | 114 | ||
| X89569.1 | - | 4 - 5 | 344 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abcd3
Genomic Map