Assay Details
| Gene Symbol: Abcd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008991.2 | NP_033017.2 | 2 - 3 | 246 | 65 | |
| GenBank mRNA | AK004660.1 | - | 2 - 3 | 227 | 65 | |
| AK144060.1 | - | 2 - 3 | 228 | 65 | ||
| AK153298.1 | - | 2 - 3 | 204 | 65 | ||
| BC009119.1 | - | 2 - 3 | 238 | 3154529 | 65 | |
| BC054446.1 | - | 2 - 3 | 194 | 6477711 | 65 | |
| BY123916.1 | - | 2 - 3 | 246 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abcd3
Genomic Map