Assay Details
| Gene Symbol: Xpo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020506.1 | NP_065252.1 | 12 - 13 | 1636 | 128 | |
| GenBank mRNA | AF145021.1 | - | 12 - 13 | 1636 | 128 | |
| AK084907.1 | - | 13 - 14 | 1821 | 128 | ||
| AK129430.1 | - | 12 - 13 | 1631 | 128 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Xpo4
Genomic Map