Assay Details
| Gene Symbol: Xpo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020506.1 | NP_065252.1 | 17 - 18 | 2622 | 161 | |
| GenBank mRNA | AF145021.1 | - | 17 - 18 | 2622 | 161 | |
| AK129430.1 | - | 17 - 18 | 2617 | 161 | ||
More Information
Set Membership: |
|

Xpo4
Genomic Map