Assay Details
| Gene Symbol: Npm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181345.3 | NP_851990.2 | 6 - 7 | 678 | 70 | |
| GenBank mRNA | AK054533.1 | - | 6 - 7 | 679 | 70 | |
| AY262112.1 | - | 6 - 7 | 678 | 70 | ||
| BC104058.1 | - | 6 - 7 | 640 | 40040361 | 70 | |
| BC104059.1 | - | 6 - 7 | 640 | 40040362 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Npm2
Genomic Map