Assay Details
| Gene Symbol: Npm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181345.3 | NP_851990.2 | 7 - 8 | 714 | 99 | |
| GenBank mRNA | AK054533.1 | - | 7 - 8 | 715 | 99 | |
| AY262112.1 | - | 7 - 8 | 714 | 99 | ||
| BC104058.1 | - | 7 - 8 | 676 | 40040361 | 99 | |
| BC104059.1 | - | 7 - 8 | 676 | 40040362 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Npm2
Genomic Map