Assay Details
| Gene Symbol: Npm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181345.3 | NP_851990.2 | 8 - 9 | 748 | 61 | |
| GenBank mRNA | AY262112.1 | - | 8 - 9 | 748 | 61 | |
| BC104058.1 | - | 8 - 9 | 710 | 40040361 | 61 | |
| BC104059.1 | - | 8 - 9 | 710 | 40040362 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
chromatin remodeling
regulation of exit from mitosis
multicellular organism development
single fertilization
embryo development
chromatin modification
positive regulation of catalytic activity
positive regulation of DNA replication
positive regulation of meiotic nuclear division
positive regulation of DNA metabolic process
protein homooligomerization
nucleic acid binding
chromatin binding
enzyme binding
chromatin remodeling
regulation of exit from mitosis
multicellular organism development
single fertilization
embryo development
chromatin modification
positive regulation of catalytic activity
positive regulation of DNA replication
positive regulation of meiotic nuclear division
positive regulation of DNA metabolic process
protein homooligomerization
nucleic acid binding
chromatin binding
enzyme binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P289679 | AK054533.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P289679 | AK054533.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 704433 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 432515 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 747901 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 648516 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 284189 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 157220 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154526 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P289679 | AK054533.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P289679 | AK054533.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P289679 | AK054533.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P289679 | AK054533.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29702682 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29702718 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29702712 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29702713 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29702730 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29702726 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29702716 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29702717 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29702709 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29702721 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29702731 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29702706 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29702729 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29702723 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29702705 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29702714 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29702685 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29702711 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29702728 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29702683 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154526 | - | - |
| Whole Mouse Genome G4122A | A_51_P289679 | AK054533.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P289679 | AK054533.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135209 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154526 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11695340 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11702177 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11695335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29702677 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11695332 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29702711 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29702683 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11695340 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11769154 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29702712 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29702701 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29702721 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29702682 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29702689 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11695328 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29702696 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11223035 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29702716 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11223037 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11677647 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29702707 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29702709 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11155448 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29702694 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29702713 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29702690 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29702685 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11245326 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11677649 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29702699 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11245320 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29702703 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11623244 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11409506 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29702705 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10421449 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10421454 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10421452 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10421453 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10421450 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10421446 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10421447 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10421451 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10421448 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10421455 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10421445 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10421444 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400018383.mm.1 | NM_181345.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400018383.mm.1 | NM_181345.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400018383.mm.1 | TC1400002348.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400018382.mm.1 | NM_181345.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400018382.mm.1 | NM_181345.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400018382.mm.1 | TC1400002348.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 301418 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6040139 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006040139 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2704678 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2704678 | - | - |
| mouse-6 v1.1 (Illumina) | scl0328440.4_21-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6040139 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0328440.4_21-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6040139 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2704678 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2704678 | - | - |

Npm2
Genomic Map