Assay Details
| Gene Symbol: Tcp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290712.1 | NP_001277641.1 | 9 - 10 | 1440 | 77 | |
| NM_013686.4 | NP_038714.2 | 10 - 11 | 1526 | 77 | ||
| GenBank mRNA | AK149611.1 | - | 10 - 11 | 1391 | 77 | |
| AK149755.1 | - | 10 - 11 | 1392 | 77 | ||
| AK150666.1 | - | 8 - 9 | 2951 | 77 | ||
| AK150744.1 | - | 8 - 9 | 2951 | 77 | ||
| AK151068.1 | - | 10 - 11 | 1391 | 77 | ||
| AK151445.1 | - | 9 - 10 | 1330 | 77 | ||
| AK151903.1 | - | 8 - 9 | 2951 | 77 | ||
| AK152633.1 | - | 8 - 9 | 2951 | 77 | ||
| AK152759.1 | - | 10 - 11 | 1391 | 77 | ||
| AK153037.1 | - | 10 - 11 | 1391 | 77 | ||
| AK153430.1 | - | 9 - 10 | 1330 | 77 | ||
| AK165665.1 | - | 10 - 11 | 1393 | 77 | ||
| AK166828.1 | - | 10 - 11 | 1390 | 77 | ||
| AK166966.1 | - | 10 - 11 | 1414 | 77 | ||
| AK167529.1 | - | 10 - 11 | 1391 | 77 | ||
| AK178327.1 | - | 2 - 3 | 249 | 77 | ||
| BC003809.1 | - | 10 - 11 | 1374 | 3599670 | 77 | |
| D90344.1 | - | 10 - 11 | 1341 | 77 | ||
| M20130.1 | - | 3 - 4 | 414 | 77 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tcp1
Genomic Map