Assay Details
| Gene Symbol: Tcp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290712.1 | NP_001277641.1 | 7 - 8 | 1123 | 57 | |
| NM_013686.4 | NP_038714.2 | 8 - 9 | 1209 | 57 | ||
| GenBank mRNA | AK149611.1 | - | 8 - 9 | 1074 | 57 | |
| AK149755.1 | - | 8 - 9 | 1075 | 57 | ||
| AK150666.1 | - | 6 - 7 | 2634 | 57 | ||
| AK150744.1 | - | 6 - 7 | 2634 | 57 | ||
| AK151068.1 | - | 8 - 9 | 1074 | 57 | ||
| AK151445.1 | - | 7 - 8 | 1013 | 57 | ||
| AK151903.1 | - | 6 - 7 | 2634 | 57 | ||
| AK152633.1 | - | 6 - 7 | 2634 | 57 | ||
| AK152641.1 | - | 8 - 9 | 1066 | 57 | ||
| AK152759.1 | - | 8 - 9 | 56 | |||
| AK153037.1 | - | 8 - 9 | 56 | |||
| AK153430.1 | - | 7 - 8 | 1013 | 57 | ||
| AK165665.1 | - | 8 - 9 | 1076 | 57 | ||
| AK166828.1 | - | 8 - 9 | 1073 | 57 | ||
| AK166966.1 | - | 8 - 9 | 1097 | 57 | ||
| AK167529.1 | - | 8 - 9 | 1074 | 57 | ||
| BC003809.1 | - | 8 - 9 | 1057 | 3599670 | 57 | |
| M12899.1 | - | 8 - 9 | 1032 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tcp1
Genomic Map