Assay Details
| Gene Symbol: Clasrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016680.5 | NP_057889.3 | 1 - 2 | 74 | 62 | |
| NR_125832.1 | 1 - 2 | 74 | 62 | |||
| NR_125833.1 | 1 - 2 | 74 | 62 | |||
| NR_125834.1 | 1 - 2 | 74 | 62 | |||
| GenBank mRNA | AK159443.1 | - | 1 - 2 | 43 | 62 | |
| BC051916.1 | - | 1 - 2 | 49 | 5707517 | 62 | |
| BC060092.1 | - | 1 - 2 | 47 | 62 | ||
| BC079543.1 | - | 1 - 2 | 37 | 62 | ||
| BC106129.1 | - | 1 - 2 | 53 | 62 | ||
| CJ173924.1 | - | 1 - 2 | 56 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC051916.1, BC106129.1, NM_016680.5 transcripts.
Set Membership: |
|

Clasrp
Genomic Map