Assay Details
| Gene Symbol: Clasrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016680.5 | NP_057889.3 | 7 - 8 | 705 | 70 | |
| NR_125832.1 | 7 - 8 | 705 | 70 | |||
| NR_125834.1 | 7 - 8 | 705 | 70 | |||
| NR_125835.1 | 7 - 8 | 667 | 70 | |||
| NR_125836.1 | 5 - 6 | 569 | 70 | |||
| GenBank mRNA | AB080582.1 | - | 6 - 7 | 568 | 70 | |
| AB080583.1 | - | 6 - 7 | 568 | 70 | ||
| AF042799.1 | - | 7 - 8 | 670 | 70 | ||
| AK009334.1 | - | 7 - 8 | 656 | 70 | ||
| AK159443.1 | - | 7 - 8 | 674 | 70 | ||
| BC010728.1 | - | 7 - 8 | 656 | 70 | ||
| BC051916.1 | - | 7 - 8 | 680 | 5707517 | 70 | |
| BC060092.1 | - | 6 - 7 | 3073 | 70 | ||
| BC079543.1 | - | 6 - 7 | 3063 | 70 | ||
| BC103781.1 | - | 5 - 6 | 535 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Clasrp
Genomic Map