Assay Details
| Gene Symbol: Mark4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172279.1 | NP_758483.1 | 15 - 16 | 1884 | 104 | |
| GenBank mRNA | AK035668.1 | - | 3 - 4 | 551 | 104 | |
| AK122565.2 | - | 11 - 12 | 1530 | 104 | ||
| AK146784.1 | - | 16 - 17 | 2219 | 104 | ||
| AY151083.1 | - | 15 - 16 | 1884 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK035668.1 transcripts.
Set Membership: |
|

Mark4
Genomic Map