Assay Details
| Gene Symbol: Mark4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172279.1 | NP_758483.1 | 7 - 8 | 549 | 74 | |
| GenBank mRNA | AK122565.2 | - | 3 - 4 | 195 | 74 | |
| AK146784.1 | - | 8 - 9 | 884 | 74 | ||
| AY151083.1 | - | 7 - 8 | 549 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mark4
Genomic Map