Assay Details
| Gene Symbol: Ppp1r13l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001010836.3 | NP_001010836.1 | 8 - 9 | 1927 | 155 | |
| GenBank mRNA | AK146098.1 | - | 8 - 9 | 1905 | 155 | |
| AK170535.1 | - | 8 - 9 | 1928 | 155 | ||
| AY869711.1 | - | 8 - 9 | 1897 | 155 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1r13l
Genomic Map