Assay Details
| Gene Symbol: Gulo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178747.3 | NP_848862.1 | 3 - 4 | 390 | 79 | |
| GenBank mRNA | AK077740.1 | - | 3 - 4 | 391 | 79 | |
| AK167460.1 | - | 3 - 4 | 379 | 79 | ||
| AY453064.1 | - | 2 - 3 | 227 | 79 | ||
| BC019856.1 | - | 3 - 4 | 337 | 5123684 | 79 | |
| BC028822.1 | - | 3 - 4 | 346 | 5097681 | 79 | |
| BC028828.1 | - | 3 - 4 | 349 | 5101228 | 79 | |
| BC034835.1 | - | 2 - 3 | 146 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gulo
Genomic Map