Assay Details
| Gene Symbol: Esco2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028039.2 | NP_082315.3 | 5 - 6 | 1185 | 83 | |
| GenBank mRNA | AA189671.1 | - | 3 - 4 | 257 | 83 | |
| AK144685.1 | - | 5 - 6 | 1141 | 83 | ||
| BC071275.1 | - | 5 - 6 | 1118 | 6331551 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Esco2
Genomic Map