Assay Details
| Gene Symbol: Akap11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164503.1 | NP_001157975.1 | 6 - 7 | 819 | 78 | |
| GenBank mRNA | AK158300.1 | - | 6 - 7 | 740 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Akap11
Genomic Map