Assay Details
| Gene Symbol: Akap11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164503.1 | NP_001157975.1 | 9 - 10 | 5509 | 96 | |
| GenBank mRNA | AK032821.1 | - | 3 - 4 | 1189 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Akap11
Genomic Map