Assay Details
| Gene Symbol: Lamb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277928.1 | NP_001264857.1 | 18 - 19 | 2832 | 81 | |
| NM_008484.2 | NP_032510.2 | 18 - 19 | 2871 | 81 | ||
| GenBank mRNA | AK078970.1 | - | 3 - 4 | 387 | 81 | |
| AK155769.1 | - | 3684 | 81 | |||
| BC008516.1 | - | 18 - 19 | 2832 | 3592888 | 81 | |
| U43298.1 | - | 18 - 19 | 2847 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lamb3
Genomic Map