Assay Details
| Gene Symbol: Olfr297 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146618.2 | NP_666829.2 | 1 | 761 | 82 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2001415 | NM_146618.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2001415 | NM_146618.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P286278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25294995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25295019 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25294994 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25295013 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25295016 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25295024 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25294998 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25295035 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25295010 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25295020 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25295002 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25295005 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25294988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25295014 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25295023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25295015 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25294993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25295034 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25295001 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25295032 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25295009 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25294991 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25295000 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25294989 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25295021 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25295008 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25294996 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25295018 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25294990 | - | - |
| Whole Mouse Genome G4122A | A_52_P286278 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123995 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06636922 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06636915 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06636930 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06636928 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06636909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25294975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25294991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25294974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25295032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25294979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25295020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25295002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25295040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25295034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25295038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25294986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25295009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25294998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25295001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25294989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25295008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25295019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25294987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25294988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25294977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25295024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25294980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25295018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25294996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25295013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25294994 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06407504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25294986 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25294995 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25295015 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25295035 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25295030 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25294975 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25295008 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25295032 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25294984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25295001 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25295034 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25295019 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25295002 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25294996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25294985 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25295000 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25294976 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25295018 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25295005 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25295038 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25295023 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06246478 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25295009 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25294994 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25294998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25295014 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25294979 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25294981 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25295016 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25294977 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25295021 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25294991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25294989 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25294988 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25295013 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06636907 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25295024 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10554728 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10554727 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 733055 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3360400 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003360400 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1252914 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1252914 | - | - |
| mouse-6 v1.1 (Illumina) | scl0258611.1_186-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3360400 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0258611.1_186-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3360400 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1252914 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1252914 | - | - |

Olfr297
Genomic Map