Assay Details
| Gene Symbol: Ap5m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144535.4 | NP_653118.3 | 4 - 5 | 1390 | 159 | |
| GenBank mRNA | AK030928.1 | - | 4 - 5 | 1317 | 159 | |
| AK031527.1 | - | 4 - 5 | 1789 | 159 | ||
| AK041400.1 | - | 4 - 5 | 891 | 159 | ||
| AK051372.1 | - | 4 - 5 | 1337 | 159 | ||
| AK154166.1 | - | 4 - 5 | 1343 | 159 | ||
| AK164148.1 | - | 4 - 5 | 1161 | 159 | ||
| AK170232.1 | - | 4 - 5 | 1167 | 159 | ||
| BC023094.1 | - | 4 - 5 | 1390 | 4017596 | 159 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ap5m1
Genomic Map