Assay Details
| Gene Symbol: Naa30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015640.1 | - | 2 - 3 | 907 | 110 | |
| AK044235.1 | - | 1 - 2 | 554 | 110 | ||
| BC157931.1 | - | 2 - 3 | 827 | 9088104 | 110 | |
| BC158065.1 | - | 2 - 3 | 827 | 9007364 | 110 | |
| BC172029.1 | - | 2 - 3 | 827 | 9054315 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|

Naa30
Genomic Map