Assay Details
| Gene Symbol: Naa30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081430.1 | NP_001074899.1 | 4 - 5 | 1108 | 149 | |
| GenBank mRNA | AK015640.1 | - | 4 - 5 | 1081 | 149 | |
| AK044235.1 | - | 3 - 4 | 728 | 149 | ||
| BC054060.1 | - | 2 - 3 | 335 | 149 | ||
| BC157931.1 | - | 4 - 5 | 1001 | 9088104 | 149 | |
| BC158065.1 | - | 4 - 5 | 1001 | 9007364 | 149 | |
| BC172029.1 | - | 4 - 5 | 1001 | 9054315 | 149 | |
More Information
Updated Mapping Information
Set Membership: |
|

Naa30
Genomic Map