Assay Details
| Gene Symbol: Mettl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029990.1 | NP_001025161.1 | 10 - 11 | 76 | ||
| GenBank mRNA | AK009576.1 | - | 10 - 11 | 76 | ||
| AK032357.1 | - | 76 | ||||
| AK155096.1 | - | 10 - 11 | 76 | |||
| AK156895.1 | - | 9 - 10 | 76 | |||
| AK172330.1 | - | 9 - 10 | 76 | |||
| BC138944.1 | - | 10 - 11 | 8861966 | 76 | ||
| BC145472.1 | - | 9 - 10 | 76 | |||
| BC145473.1 | - | 8 - 9 | 76 | |||
| BC145474.1 | - | 9 - 10 | 76 | |||
| CO043212.1 | - | 4 - 5 | 76 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mettl17
Genomic Map