Assay Details
| Gene Symbol: Mettl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029990.1 | NP_001025161.1 | 4 - 5 | 550 | 71 | |
| GenBank mRNA | AK009576.1 | - | 4 - 5 | 464 | 71 | |
| AK032357.1 | - | 541 | 71 | |||
| AK087101.1 | - | 4 - 5 | 482 | 71 | ||
| AK155096.1 | - | 4 - 5 | 469 | 71 | ||
| AK156895.1 | - | 3 - 4 | 600 | 71 | ||
| AK172330.1 | - | 3 - 4 | 600 | 71 | ||
| BC138944.1 | - | 4 - 5 | 503 | 8861966 | 71 | |
| BC145472.1 | - | 4 - 5 | 503 | 71 | ||
| BC145473.1 | - | 3 - 4 | 586 | 71 | ||
| BC145474.1 | - | 4 - 5 | 503 | 71 | ||
| CF172670.1 | - | 4 - 5 | 550 | 71 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mettl17
Genomic Map