Assay Details
| Gene Symbol: Mettl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029990.1 | NP_001025161.1 | 9 - 10 | 982 | 66 | |
| GenBank mRNA | AK009576.1 | - | 9 - 10 | 895 | 66 | |
| AK032357.1 | - | 1594 | 66 | |||
| AK155096.1 | - | 9 - 10 | 901 | 66 | ||
| AK156895.1 | - | 8 - 9 | 1212 | 66 | ||
| AK172330.1 | - | 8 - 9 | 1212 | 66 | ||
| BC138944.1 | - | 9 - 10 | 935 | 8861966 | 66 | |
| BC145472.1 | - | 8 - 9 | 864 | 66 | ||
| BC145473.1 | - | 7 - 8 | 947 | 66 | ||
| BC145474.1 | - | 8 - 9 | 864 | 66 | ||
| CO043212.1 | - | 5 - 6 | 66 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mettl17
Genomic Map