Assay Details
| Gene Symbol: Tppp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128634.1 | NP_001122106.1 | 1 - 2 | 196 | 76 | |
| GenBank mRNA | BC049709.1 | - | 2 - 3 | 310 | 6740792 | 76 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tppp2
Genomic Map