Assay Details
| Gene Symbol: Slc22a17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB012808.1 | - | 3 - 4 | 442 | 55 | |
| AK163116.1 | - | 1 - 2 | 865 | 55 | ||
| BC005727.1 | - | 2 - 3 | 722 | 55 | ||
| BC062878.1 | - | 2 - 3 | 518 | 5696414 | 55 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB012808.1 transcripts.
Set Membership: |
|

Slc22a17
Genomic Map