Assay Details
| Gene Symbol: Slc22a17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK163116.1 | - | 2 - 3 | 966 | 65 | |
| BC005727.1 | - | 3 - 4 | 823 | 65 | ||
| BC062878.1 | - | 3 - 4 | 619 | 5696414 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc22a17
Genomic Map