Assay Details
| Gene Symbol: Slc22a17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021551.4 | NP_067526.2 | 4 - 5 | 1080 | 60 | |
| GenBank mRNA | AB012808.1 | - | 6 - 7 | 866 | 60 | |
| AK007059.1 | - | 4 - 5 | 1025 | 60 | ||
| BC062878.1 | - | 5 - 6 | 942 | 5696414 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc22a17
Genomic Map